The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031648.2 | NP_113836.2 | 7 - 8 | 401 | 105 |
XM_006228838.2 | XP_006228900.1 | 8 - 9 | 621 | 105 | |
XM_006228839.2 | XP_006228901.1 | 7 - 8 | 396 | 105 | |
XM_008759184.1 | XP_008757406.1 | 7 - 8 | 410 | 105 | |
GenBank mRNA | CB712570.1 | - | 7 - 8 | 401 | 105 |
FM109156.1 | - | 7 - 8 | 355 | 105 | |
FQ211349.1 | - | 7 - 8 | 412 | 105 | |
FQ224131.1 | - | 7 - 8 | 364 | 105 | |
FQ224338.1 | - | 7 - 8 | 105 |