Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap16-5
Entrez Gene ID
680703
Gene Name
keratin associated protein 16-5
Gene Aliases
-
Chromosome Location
Chr.11: 28899792 - 28900376 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109424.1 NP_001102894.1 1 - 1 345 178

View Full Product Details