Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jtb
Entrez Gene ID
29439
Gene Name
jumping translocation breakpoint
Gene Aliases
-
Chromosome Location
Chr.2: 189591707 - 189595926 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019213.2 NP_062086.1 1 - 2 357 78
GenBank mRNA AB016489.1 - 1 - 2 411 78
BC062090.1 - 1 - 2 357 78

View Full Product Details