Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csgalnact2
Entrez Gene ID
297554
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
Gene Aliases
Galnact2, RGD1563660
Chromosome Location
Chr.4: 150153835 - 150185058 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106616.1 NP_001100086.1 7 - 8 1709 76
XM_006237181.2 XP_006237243.1 7 - 8 1482 76
GenBank mRNA FQ221326.1 - 7 - 8 1621 76

View Full Product Details