Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem72
Entrez Gene ID
362424
Gene Name
transmembrane protein 72
Gene Aliases
RGD1308850
Chromosome Location
Chr.4: 148819906 - 148845199 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108643.1 NP_001102113.1 4 - 5 690 62

View Full Product Details