The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106627.3 | NP_001100097.2 | 8 - 9 | 737 | 67 |
XM_008763378.1 | XP_008761600.1 | 4 - 5 | 1174 | 67 | |
GenBank mRNA | BC160907.1 | - | 8 - 9 | 737 | 67 |