Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss32
Entrez Gene ID
302970
Gene Name
protease, serine, 32
Gene Aliases
-
Chromosome Location
Chr.10: 13196700 - 13202783 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106983.1 NP_001100453.1 1 - 2 377 81

View Full Product Details