Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gfap
Entrez Gene ID
24387
Gene Name
glial fibrillary acidic protein
Gene Aliases
-
Chromosome Location
Chr.10: 90990762 - 90999501 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017009.2 NP_058705.2 5 - 6 897 81
XM_008768261.1 XP_008766483.1 5 - 6 963 81
GenBank mRNA BC088851.1 - 5 - 6 897 81
EF094476.1 - 5 - 6 968 81
EF094477.1 - 1 - 2 115 81
L27219.1 - 5 - 6 901 81
U03700.1 - 5 - 6 921 81

View Full Product Details