Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmvk
Entrez Gene ID
310645
Gene Name
phosphomevalonate kinase
Gene Aliases
-
Chromosome Location
Chr.2: 188784329 - 188793885 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008352.1 NP_001008353.1 4 - 5 485 71
GenBank mRNA BC086349.1 - 4 - 5 485 71
FQ220630.1 - 4 - 5 535 71
FQ226470.1 - 4 - 5 615 71
FQ233923.1 - 4 - 5 615 71
FQ234368.1 - 4 - 5 615 71

View Full Product Details