The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138839.2 | NP_620194.1 | 11 - 12 | 1186 | 85 |
XM_006247078.2 | XP_006247140.1 | 11 - 12 | 1664 | 85 | |
XM_006247079.2 | XP_006247141.1 | 12 - 13 | 1208 | 85 | |
GenBank mRNA | AF411216.1 | - | 11 - 12 | 1132 | 85 |
BC061721.1 | - | 11 - 12 | 1186 | 85 | |
FQ220200.1 | - | 2 - 3 | 338 | 85 | |
FQ220288.1 | - | 2 - 3 | 352 | 85 | |
FQ220745.1 | - | 1 - 2 | 307 | 85 | |
FQ220965.1 | - | 2 - 3 | 338 | 85 |