The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039163.1 | NP_001034252.1 | 2 - 3 | 962 | 58 |
GenBank mRNA | AB218813.1 | - | 2 - 3 | 962 | 58 |