The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034144.1 | NP_001029316.1 | 1 - 2 | 300 | 96 |
GenBank mRNA | BC100628.1 | - | 1 - 2 | 300 | 96 |