Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spsb3
Entrez Gene ID
302981
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
Tce1
Chromosome Location
Chr.10: 14248710 - 14253815 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106988.1 NP_001100458.1 4 - 5 914 88

View Full Product Details