The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032618.1 | NP_116007.1 | 2 - 3 | 432 | 119 |
GenBank mRNA | AF223951.1 | - | 2 - 3 | 416 | 119 |
FM107575.1 | - | 2 - 3 | 432 | 119 |