The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017376.1 | NP_001017376.1 | - | 752 | 150 |
XM_006252293.2 | XP_006252355.1 | 4 - 5 | 1258 | 150 | |
GenBank mRNA | BC092627.1 | - | - | 752 | 150 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006252260.2 | XP_006252322.1 | 4 - 5 | 1258 | 150 |
XM_008770837.1 | XP_008769059.1 | 4 - 5 | 1297 | 150 |