Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srpx2
Entrez Gene ID
317181
Gene Name
sushi-repeat-containing protein, X-linked 2
Gene Aliases
RGD1562444
Chromosome Location
Chr.X: 104734368 - 104760658 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108243.2 NP_001101713.1 4 - 5 429 139
GenBank mRNA BC168975.1 - 4 - 5 429 139

View Full Product Details