Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btk
Entrez Gene ID
367901
Gene Name
Bruton agammaglobulinemia tyrosine kinase
Gene Aliases
-
Chromosome Location
Chr.X: 105360922 - 105390580 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007798.1 NP_001007799.1 5 - 6 589 126
GenBank mRNA AY787790.1 - 5 - 6 589 126

View Full Product Details