The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106733.1 | NP_001100203.1 | 5 - 6 | 627 | 112 |
GenBank mRNA | FQ230024.1 | - | 5 - 6 | 390 | 112 |