Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkar2b
Entrez Gene ID
24679
Gene Name
protein kinase, cAMP dependent regulatory, type II beta
Gene Aliases
RATDNA
Chromosome Location
Chr.6: 51265509 - 51356383 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030020.1 NP_001025191.1 3 - 4 390 99
GenBank mRNA M12492.1 - 3 - 4 384 99

View Full Product Details