The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024892.1 | NP_001020063.1 | 3 - 4 | 374 | 150 |
XM_006257149.2 | XP_006257211.1 | 4 - 5 | 569 | 150 | |
GenBank mRNA | BC097491.1 | - | 3 - 4 | 374 | 150 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003752081.3 | XP_003752129.1 | - | 457 | 150 |
XM_006257141.2 | XP_006257203.1 | - | 569 | 150 |