Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adcy5
Entrez Gene ID
64532
Gene Name
adenylate cyclase 5
Gene Aliases
-
Chromosome Location
Chr.11: 68695839 - 68842493 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022600.1 NP_072122.1 7 - 8 2380 77
XM_008768744.1 XP_008766966.1 7 - 8 2553 77
GenBank mRNA M96159.1 - 7 - 8 2380 77

View Full Product Details