Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mphosph10
Entrez Gene ID
293828
Gene Name
M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein)
Gene Aliases
-
Chromosome Location
Chr.1: 125212869 - 125229060 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106340.1 NP_001099810.1 3 - 4 959 84
GenBank mRNA BC161909.1 - 3 - 4 932 84

View Full Product Details