The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008510.1 | NP_001008510.1 | 2 - 3 | 460 | 84 |
GenBank mRNA | BC086557.1 | - | 2 - 3 | 460 | 84 |
FQ233960.1 | - | 3 - 4 | 517 | 84 | |
FQ234031.1 | - | - | 527 | 84 | |
FQ234316.1 | - | 2 - 3 | 513 | 84 |