The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014161.1 | NP_001014183.1 | 10 - 11 | 1300 | 60 |
GenBank mRNA | BC076398.1 | - | 10 - 11 | 1300 | 60 |