Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc1a1
Entrez Gene ID
25550
Gene Name
solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1
Gene Aliases
Eaac1, Eaat3, REAAC1
Chromosome Location
Chr.1: 246955017 - 247035159 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013032.3 NP_037164.3 7 - 8 859 154
GenBank mRNA BC061743.1 - 7 - 8 859 154
D63772.1 - - 890 154
U21104.1 - 7 - 8 871 154
U39555.1 - 7 - 8 949 154
X94255.1 - 7 - 8 916 154

View Full Product Details