Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc10b
Entrez Gene ID
309208
Gene Name
leucine rich repeat containing 10B
Gene Aliases
RGD1564983
Chromosome Location
Chr.1: 226500692 - 226501920 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107577.1 NP_001101047.1 1 - 1 1137 91

View Full Product Details