Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam53b
Entrez Gene ID
309060
Gene Name
family with sequence similarity 53, member B
Gene Aliases
RGD1310799
Chromosome Location
Chr.1: 204714468 - 204805738 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107556.1 NP_001101026.1 3 - 4 535 129
XM_008759886.1 XP_008758108.1 2 - 3 2415 129
XM_008759887.1 XP_008758109.1 2 - 3 4343 129
XM_008759888.1 XP_008758110.1 2 - 3 4304 129

View Full Product Details