The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107157.1 | NP_001100627.1 | 4 - 5 | 718 | 109 |
XM_006249158.1 | XP_006249220.1 | 3 - 4 | 664 | 109 | |
XM_006249159.1 | XP_006249221.1 | 3 - 4 | 664 | 109 | |
XM_008769137.1 | XP_008767359.1 | 3 - 4 | 519 | 109 | |
GenBank mRNA | AY724523.1 | - | 2 - 3 | 499 | 109 |
BC166738.1 | - | 3 - 4 | 605 | 109 |