The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025718.1 | NP_001020889.1 | 2 - 3 | 164 | 74 |
XM_006253451.2 | XP_006253513.1 | 3 - 4 | 429 | 74 | |
XM_008771397.1 | XP_008769619.1 | 4 - 5 | 432 | 74 | |
XM_008771398.1 | XP_008769620.1 | 4 - 5 | 511 | 74 | |
XM_008771399.1 | XP_008769621.1 | 4 - 5 | 339 | 74 | |
XM_008771400.1 | XP_008769622.1 | 2 - 3 | 362 | 74 | |
GenBank mRNA | BC098789.1 | - | 2 - 3 | 164 | 74 |