The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025718.1 | NP_001020889.1 | 1 - 2 | 55 | 91 |
XM_008771400.1 | XP_008769622.1 | 1 - 2 | 253 | 91 | |
GenBank mRNA | BC098789.1 | - | 1 - 2 | 55 | 91 |