Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc41a1
Entrez Gene ID
363985
Gene Name
solute carrier family 41 (magnesium transporter), member 1
Gene Aliases
-
Chromosome Location
Chr.13: 48607308 - 48627999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108855.1 NP_001102325.1 9 - 10 2235 106
XM_008769486.1 XP_008767708.1 8 - 9 1358 106

View Full Product Details