The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013119.1 | NP_037251.1 | 23 - 24 | 4554 | 107 |
XM_008761914.1 | XP_008760136.1 | 22 - 23 | 4453 | 107 | |
XM_008761915.1 | XP_008760137.1 | 23 - 24 | 4456 | 107 | |
XM_008761916.1 | XP_008760138.1 | 22 - 23 | 4454 | 107 | |
XM_008761917.1 | XP_008760139.1 | 22 - 23 | 4453 | 107 | |
XM_008761918.1 | XP_008760140.1 | 22 - 23 | 4366 | 107 | |
XM_008761919.1 | XP_008760141.1 | 22 - 23 | 4357 | 107 | |
XM_008761920.1 | XP_008760142.1 | 22 - 23 | 4357 | 107 | |
GenBank mRNA | Y00766.1 | - | 23 - 24 | 4554 | 107 |