Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nutm1
Entrez Gene ID
366153
Gene Name
NUT midline carcinoma, family member 1
Gene Aliases
Nut, RGD1564529
Chromosome Location
Chr.3: 103733935 - 103745236 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108957.1 NP_001102427.1 3 - 4 957 69

View Full Product Details