Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc38a11
Entrez Gene ID
362141
Gene Name
solute carrier family 38, member 11
Gene Aliases
RGD1306005
Chromosome Location
Chr.3: 51370388 - 51416875 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_342441.7 XP_342442.5 9 - 10 1080 76

View Full Product Details