Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otud1
Entrez Gene ID
498803
Gene Name
OTU deubiquitinase 1
Gene Aliases
RGD1563344
Chromosome Location
Chr.17: 86406647 - 86410670 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_574086.6 XP_574086.3 1 - 1 3075 79

View Full Product Details