Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phyh
Entrez Gene ID
114209
Gene Name
phytanoyl-CoA 2-hydroxylase
Gene Aliases
-
Chromosome Location
Chr.17: 77287580 - 77304482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053674.1 NP_446126.1 2 - 3 143 69
GenBank mRNA AF121345.1 - 2 - 3 143 69
BC086573.1 - 2 - 3 138 69

View Full Product Details