The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106125.1 | NP_001099595.1 | 10 - 11 | 1452 | 85 |
XM_006254307.2 | XP_006254369.1 | 11 - 12 | 5366 | 85 |