Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Msr1
Entrez Gene ID
498638
Gene Name
macrophage scavenger receptor 1
Gene Aliases
RGD1564316
Chromosome Location
Chr.16: 56817714 - 56900024 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191939.1 NP_001178868.1 2 - 3 219 91
XM_006253206.2 XP_006253268.1 3 - 4 408 91
XM_006253207.2 XP_006253269.1 3 - 4 403 91

View Full Product Details