Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Neil3
Entrez Gene ID
290729
Gene Name
nei endonuclease VIII-like 3 (E. coli)
Gene Aliases
RGD1310562
Chromosome Location
Chr.16: 41079444 - 41138505 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170346.1 NP_001163817.1 10 - 11 1752 54

View Full Product Details