Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a17
Entrez Gene ID
305886
Gene Name
solute carrier family 22, member 17
Gene Aliases
24p3R, Boct, rBOCT
Chromosome Location
Chr.15: 33569824 - 33576972 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177421.3 NP_803156.2 3 - 4 827 67
XM_006251965.1 XP_006252027.1 3 - 4 1465 67
GenBank mRNA AB040056.1 - 3 - 4 834 67
BC099072.1 - 3 - 4 827 67
FQ212273.1 - 3 - 4 829 67

View Full Product Details