Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irf9
Entrez Gene ID
305896
Gene Name
interferon regulatory factor 9
Gene Aliases
Isgf3g
Chromosome Location
Chr.15: 34282936 - 34288981 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012041.1 NP_001012041.1 4 - 5 455 77
XM_006251974.2 XP_006252036.1 4 - 5 583 77
XM_006251975.1 XP_006252037.1 4 - 5 527 77
XM_008770679.1 XP_008768901.1 4 - 5 568 77
GenBank mRNA BC079454.1 - 4 - 5 455 77

View Full Product Details