The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133311.1 | NP_579845.1 | 3 - 4 | 466 | 71 |
XM_008769421.1 | XP_008767643.1 | 3 - 4 | 618 | 71 | |
GenBank mRNA | AF269251.1 | - | 3 - 4 | 466 | 71 |