Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Seh1l
Entrez Gene ID
307347
Gene Name
SEH1-like (S. cerevisiae)
Gene Aliases
Da1-6
Chromosome Location
Chr.18: 63572088 - 63656302 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY325252.1 - - 2777 72

View Full Product Details