The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013069.2 | NP_037201.1 | 7 - 8 | 737 | 55 |
XM_006254761.2 | XP_006254823.1 | 8 - 9 | 987 | 55 | |
GenBank mRNA | BC059152.1 | - | 7 - 8 | 718 | 55 |
FQ219430.1 | - | 7 - 8 | 56 | ||
FQ225098.1 | - | 7 - 8 | 712 | 55 | |
FQ225666.1 | - | 5 - 6 | 356 | 55 | |
FQ227455.1 | - | 7 - 8 | 717 | 55 | |
FQ227561.1 | - | 7 - 8 | 721 | 55 | |
FQ228116.1 | - | 7 - 8 | 720 | 55 | |
FQ231756.1 | - | 7 - 8 | 721 | 55 | |
FQ232412.1 | - | 7 - 8 | 762 | 55 | |
FQ232423.1 | - | 7 - 8 | 751 | 55 | |
FQ232867.1 | - | 7 - 8 | 721 | 55 | |
FQ233706.1 | - | 7 - 8 | 720 | 55 | |
FQ234457.1 | - | 7 - 8 | 723 | 55 | |
X13044.1 | - | 7 - 8 | 693 | 55 | |
X14254.1 | - | 8 - 9 | 893 | 55 |