Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mlf2
Entrez Gene ID
312709
Gene Name
myeloid leukemia factor 2
Gene Aliases
-
Chromosome Location
Chr.4: 157452579 - 157457254 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107889.1 NP_001101359.1 7 - 8 1015 78
XM_006237363.2 XP_006237425.1 8 - 9 1005 78

View Full Product Details