Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nrip2
Entrez Gene ID
689619
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.4: 161719431 - 161729673 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001071392.5 XP_001071392.2 5 - 6 1669 95

View Full Product Details