Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrriq4
Entrez Gene ID
100363341
Gene Name
leucine-rich repeats and IQ motif containing 4
Gene Aliases
-
Chromosome Location
Chr.2: 116347120 - 116372930 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760970.1 XP_008759192.1 3 - 4 1511 138
GenBank mRNA BC168195.1 - 2 - 3 1361 138

View Full Product Details