Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc7a11
Entrez Gene ID
310392
Gene Name
solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11
Gene Aliases
-
Chromosome Location
Chr.2: 139453774 - 139528482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107673.2 NP_001101143.2 3 - 4 651 136

View Full Product Details