Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx1a
Entrez Gene ID
116470
Gene Name
syntaxin 1A (brain)
Gene Aliases
-
Chromosome Location
Chr.12: 24682046 - 24710047 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053788.2 NP_446240.2 7 - 8 578 82
GenBank mRNA AF217191.1 - 7 - 8 547 82
D10392.1 - 7 - 8 578 82
D12519.1 - 7 - 8 538 82
M95734.1 - 7 - 8 539 82

View Full Product Details