Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpst1
Entrez Gene ID
288617
Gene Name
tyrosylprotein sulfotransferase 1
Gene Aliases
-
Chromosome Location
Chr.12: 30045753 - 30112380 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011903.2 NP_001011903.2 4 - 5 1342 98
XM_008769133.1 XP_008767355.1 5 - 6 1356 98
GenBank mRNA BC078928.1 - 5 - 6 1280 98
BC105826.1 - 4 - 5 1342 98

View Full Product Details